Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PAT1 homolog 2 | PATL2 |
Pat1a, hPat1a |
-1.4kb | Anti-sense | Chr. 15 - 44665732 - 44711389 on NCBI Build GRCh38 |
| Downstream | ring finger protein 4 pseudogene | LOC100419583 | 13.8kb | Sense | Chr. 15 - 44728988 - 44732851 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map