Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724552 | LOC102724552 | -370.9kb | Anti-sense | Chr. 1 - 8424645 - 8435016 on NCBI Build GRCh38 | |
| Downstream | enolase 1 | ENO1 |
ENO1L1, HEL-S-17, MPB1, NNE, PPH |
52.7kb | Sense | Chr. 1 - 8861000 - 8879092 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map