Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | survival associated mitochondrial melanoma specific oncogenic non-coding RNA | SAMMSON | LINC01212 | -1.1mb | Anti-sense | Chr. 3 - 69999744 - 70015301 on NCBI Build GRCh38 |
| Downstream | FOXP1 antisense RNA 1 | FOXP1-AS1 | 136.3kb | Anti-sense | Chr. 3 - 71289758 - 71305854 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map