Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
UTP4, small subunit processome component |
UTP4 |
CIRH1A, CIRHIN, NAIC, TEX292 |
-20.3kb | Sense | Chr. 16 - 69132596 - 69169034 on NCBI Build GRCh38 |
| Downstream | vacuolar protein sorting 4 homolog A | VPS4A |
SKD1, SKD1A, SKD2, VPS4, VPS4-1 |
120.2kb | Sense | Chr. 16 - 69311384 - 69325043 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map