Assay Details
| Gene Symbol: IDI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011519457.1 | XP_011517759.1 | 1 | 945 | 83 | |
| XM_017016191.1 | XP_016871680.1 | 1 | 945 | 83 | ||
| GenBank mRNA | AK022980.1 | - | 1 | 1552 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | IDI2 antisense RNA 1 | IDI2-AS1 |
C10orf110, HT009, IDI2-AS |
-9.5kb | Anti-sense | Chr. 10 - 1022637 - 1044201 on NCBI Build GRCh38 |
| Downstream | WD repeat domain 37 | WDR37 | 713.0 | Anti-sense | Chr. 10 - 1056836 - 1132297 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011519457.1, XM_017016191.1 transcripts.
Set Membership: |
|

IDI1
Genomic Map