Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929486 | LOC101929486 | -160.0kb | Anti-sense | Chr. 20 - 12865204 - 12952519 on NCBI Build GRCh38 | |
| Downstream |
isthmin 1, angiogenesis inhibitor |
ISM1 |
C20orf82, ISM, Isthmin, bA149I18.1, dJ1077I2.1 |
107.1kb | Sense | Chr. 20 - 13221771 - 13326734 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map