Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6081 | MIR6081 | hsa-mir-6081 | -6.4kb | Sense | Chr. 9 - 95065350 - 95065446 on NCBI Build GRCh38 |
| Downstream | microRNA 23b | MIR23B |
MIRN23B, hsa-mir-23b, miRNA23B, mir-23b |
11.8kb | Sense | Chr. 9 - 95085208 - 95085304 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map