Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein L17 pseudogene 10 | RPL17P10 | RPL17_2_309 | -5.7kb | Sense | Chr. 2 - 201102493 - 201102990 on NCBI Build GRCh38 |
| Downstream | CASP8 and FADD like apoptosis regulator | CFLAR |
CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS |
4.8kb | Anti-sense | Chr. 2 - 201116015 - 201172688 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map