Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PAT1 homolog 2 | PATL2 |
Pat1a, hPat1a |
Sense | Chr. 15 - 44665732 - 44711389 on NCBI Build GRCh38 | |
| Downstream | beta-2-microglobulin | B2M | IMD43 | 27.3kb | Anti-sense | Chr. 15 - 44711487 - 44718159 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map