Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleus accumbens associated 1 | NACC1 |
BEND8, BTBD14B, BTBD30, NAC-1, NAC1 |
Sense | Chr. 19 - 13116848 - 13141147 on NCBI Build GRCh38 | |
| Downstream | syntaxin 10 | STX10 |
SYN10, hsyn10 |
11.7kb | Anti-sense | Chr. 19 - 13144058 - 13150374 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map