Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RAB GTPase activating protein 1 like | RABGAP1L |
HHL, TBC1D18 |
Sense | Chr. 1 - 174159414 - 174995308 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101928696 | LOC101928696 | 459.2kb | Anti-sense | Chr. 1 - 174934947 - 174954261 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map