Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928674 | LOC101928674 | -48.9kb | Sense | Chr. 17 - 78360448 - 78373909 on NCBI Build GRCh38 | |
| Downstream | DNAH17 antisense RNA 1 | DNAH17-AS1 | 56.9kb | Sense | Chr. 17 - 78484910 - 78503056 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map