Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | general transcription factor IIA subunit 2 | GTF2A2 |
HsT18745, T18745, TF2A2, TFIIA, TFIIA-12, TFIIA-gamma, TFIIAS |
-22.6kb | Sense | Chr. 15 - 59638062 - 59657541 on NCBI Build GRCh38 |
| Downstream | forkhead box B1 | FOXB1 |
FKH5, HFKH-5 |
322.2kb | Anti-sense | Chr. 15 - 60004222 - 60005943 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map