Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1 |
ARAP1 | CENTD2 | -9.6kb | Sense | Chr. 11 - 72685069 - 72752389 on NCBI Build GRCh38 |
| Downstream | microRNA 4692 | MIR4692 | 19.6kb | Anti-sense | Chr. 11 - 72783530 - 72783592 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map