Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 856 | LINC00856 | -5.4kb | Sense | Chr. 10 - 78248625 - 78252645 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 595 | LINC00595 | C10orf101 | 7.2kb | Sense | Chr. 10 - 78267328 - 78280213 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map