Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7 |
HSD3B7 |
CBAS1, PFIC4, SDR11E3 |
Anti-sense | Chr. 16 - 30985189 - 30989152 on NCBI Build GRCh38 | |
| Downstream | syntaxin 1B | STX1B |
GEFSP9, STX1B1, STX1B2 |
986.0 | Sense | Chr. 16 - 30989256 - 31010638 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map