Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome X open reading frame 38 | CXorf38 | -14.6kb | Sense | Chr. X - 40626921 - 40647678 on NCBI Build GRCh38 | |
| Downstream | MED14 opposite strand | MED14OS | MED14-AS1 | 72.5kb | Anti-sense | Chr. X - 40735396 - 40738701 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map