Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin conjugating enzyme E2 V2 | UBE2V2 |
DDVIT1, DDVit-1, EDAF-1, EDPF-1, EDPF1, MMS2, UEV-2, UEV2 |
-363.6kb | Anti-sense | Chr. 8 - 47997527 - 48063545 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101929268 | LOC101929268 | 120.5kb | Anti-sense | Chr. 8 - 48551567 - 48698510 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map