Assay Details
| Gene Symbol: CFHR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC020680.1 | - | 1 | 851 | 124 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | complement factor H | CFH |
AHUS1, AMBP1, ARMD4, ARMS1, CFHL3, FH, FHL1, HF, HF1, HF2, HUS |
-27.3kb | Sense | Chr. 1 - 196651878 - 196747504 on NCBI Build GRCh38 |
| Downstream | complement factor H related 1 | CFHR1 |
CFHL, CFHL1, CFHL1P, CFHR1P, FHR1, H36-1, H36-2, HFL1, HFL2 |
43.7kb | Sense | Chr. 1 - 196819730 - 196832189 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

CFHR3
Genomic Map