Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | trans-golgi network vesicle protein 23 homolog A | TVP23A |
FAM18A, YDR084C |
-120.0kb | Sense | Chr. 16 - 10758034 - 10818902 on NCBI Build GRCh38 |
| Downstream | C-type lectin domain family 16 member A | CLEC16A |
Gop-1, KIAA0350 |
4.4kb | Anti-sense | Chr. 16 - 10944488 - 11193272 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map