Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosomal protein S2 pseudogene 46 | RPS2P46 | RPS2_22_1525 | -87.0kb | Anti-sense | Chr. 17 - 19445929 - 19446875 on NCBI Build GRCh38 |
| Downstream |
small nucleolar RNA, H/ACA box 59B |
SNORA59B | ACA59 | 21.9kb | Sense | Chr. 17 - 19557560 - 19557711 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map