| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008787.hg.1 |
ENST00000562749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008787.hg.1 |
ENST00000562749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008787.hg.1 |
ENST00000569906.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008787.hg.1 |
ENST00000569906.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008787.hg.1 |
NR_131982.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008787.hg.1 |
NR_131982.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159836.hg.1 |
ENST00000562749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159836.hg.1 |
NR_131982.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159836.hg.1 |
ENST00000562749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159836.hg.1 |
NR_131982.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159836.hg.1 |
TC0X00008787.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008787.hg.1 |
ENST00000562749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008787.hg.1 |
ENST00000562749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008787.hg.1 |
ENST00000569906.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008787.hg.1 |
ENST00000569906.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008787.hg.1 |
NR_131982.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008787.hg.1 |
NR_131982.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159839.hg.1 |
ENST00000562749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159839.hg.1 |
ENST00000569906.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159839.hg.1 |
NR_131982.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159839.hg.1 |
ENST00000562749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159839.hg.1 |
ENST00000569906.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00159839.hg.1 |
NR_131982.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00159839.hg.1 |
TC0X00008787.hg.1 |
Clariom D TC |