Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin conjugating enzyme E2 Z | UBE2Z |
HOYS7, USE1 |
-15.9kb | Sense | Chr. 17 - 48908369 - 48929060 on NCBI Build GRCh38 |
| Downstream | gastric inhibitory polypeptide | GIP | 12.9kb | Anti-sense | Chr. 17 - 48958556 - 48968593 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map