Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 218-2 | MIR218-2 |
MIRN218-2, mir-218-2 |
-243.9kb | Sense | Chr. 5 - 168768146 - 168768255 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC728095 | LOC728095 | 79.0 | Anti-sense | Chr. 5 - 169013227 - 169037998 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map