Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105377621 | LOC105377621 | Sense | Chr. 4 - 102418602 - 102502486 on NCBI Build GRCh38 | ||
| Downstream | nuclear factor kappa B subunit 1 | NFKB1 |
CVID12, EBP-1, KBF1, NF-kB1, NF-kappa-B, NF-kappaB, NFKB-p105, NFKB-p50, NFkappaB, p105, p50 |
10.0 | Anti-sense | Chr. 4 - 102501329 - 102617302 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map