Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | N-acetylated alpha-linked acidic dipeptidase-like 2 | NAALADL2 | Anti-sense | Chr. 3 - 174439669 - 175809510 on NCBI Build GRCh38 | ||
| Downstream | NAALADL2 antisense RNA 3 | NAALADL2-AS3 | 203.7kb | Sense | Chr. 3 - 175079312 - 175115242 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map