Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1214 | LINC01214 | -24.8kb | Anti-sense | Chr. 3 - 150265407 - 150323747 on NCBI Build GRCh38 | |
| Downstream | TSC22 domain family member 2 | TSC22D2 |
TILZ4a, TILZ4b, TILZ4c |
59.4kb | Sense | Chr. 3 - 150408268 - 150460119 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map