Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3182 | MIR3182 | mir-3182 | -226.6kb | Sense | Chr. 16 - 83508346 - 83508408 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC102724163 | LOC102724163 | 61.7kb | Anti-sense | Chr. 16 - 83797236 - 83803976 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map