Assay Details
| Gene Symbol: N4BP2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011535303.2 | XP_011533605.1 | 1 | 62 | ||
| XM_017020836.1 | XP_016876325.1 | 1 | 62 | |||
| XM_017020839.1 | XP_016876328.1 | 1 | 62 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
BRCA2, DNA repair associated |
BRCA2 |
BRCC2, BROVCA2, FACD, FAD, FAD1, FANCD, FANCD1, GLM3, PNCA2, XRCC11 |
-28.8kb | Sense | Chr. 13 - 32315480 - 32399672 on NCBI Build GRCh38 |
| Downstream | NEDD4 binding protein 2 like 2 | N4BP2L2 |
92M18.3, CG005, CG016, PFAAP5 |
3.4kb | Anti-sense | Chr. 13 - 32432488 - 32542710 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011535303.2,XM_017020836.1,XM_017020839.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

N4BP2L1
Genomic Map