Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 30c-2 | MIR30C2 |
MIRN30C2, mir-30c-2 |
Anti-sense | Chr. 6 - 71376960 - 71377031 on NCBI Build GRCh38 | |
| Downstream | microRNA 30a | MIR30A |
MIRN30A, mir-30a |
26.4kb | Anti-sense | Chr. 6 - 71403551 - 71403621 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map