Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | N4BPL2 intronic transcript 2 | N4BP2L2-IT2 | CG030 | -18.9kb | Sense | Chr. 13 - 32504506 - 32509395 on NCBI Build GRCh38 |
| Downstream | PDS5 cohesin associated factor B | PDS5B |
APRIN, AS3, CG008 |
57.7kb | Anti-sense | Chr. 13 - 32586427 - 32778020 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map