Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIB43A domain with coiled-coils 1 | RIBC1 | 2610028I09Rik | Sense | Chr. X - 53422782 - 53431120 on NCBI Build GRCh38 | |
| Downstream | hydroxysteroid 17-beta dehydrogenase 10 | HSD17B10 |
17b-HSD10, ABAD, CAMR, DUPXp11.22, ERAB, HADH2, HCD2, MHBD, MRPP2, MRX17, MRX31, MRXS10, SCHAD, SDR5C1 |
5.0kb | Anti-sense | Chr. X - 53431258 - 53434376 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map