Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mediator complex subunit 30 | MED30 |
MED30S, THRAP6, TRAP25 |
-256.1kb | Anti-sense | Chr. 8 - 117520546 - 117540262 on NCBI Build GRCh38 |
| Downstream | exostosin glycosyltransferase 1 | EXT1 |
EXT, LGCR, LGS, TRPS2, TTV |
349.0 | Sense | Chr. 8 - 117797496 - 118111819 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map