Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102724330 | LOC102724330 | -102.6kb | Sense | Chr. 8 - 51899241 - 51949886 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101929341 | LOC101929341 | 68.1kb | Sense | Chr. 8 - 52194362 - 52199552 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map