Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | leucine rich repeat containing 8 family member B | LRRC8B |
TA-LRRP, TALRRP |
-5.9kb | Anti-sense | Chr. 1 - 89522627 - 89597861 on NCBI Build GRCh38 |
| Downstream | leucine rich repeat containing 8 family member C | LRRC8C |
AD158, FAD158 |
7.8kb | Anti-sense | Chr. 1 - 89615810 - 89719535 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map