Assay Details
| Gene Symbol: GCM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017011390.1 | XP_016866879.1 | 10 | 5112 | 99 | |
| GenBank mRNA | AF075013.1 | - | 1 | 152 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | F-box protein 9 | FBXO9 |
FBX9, NY-REN-57, VCIA1, dJ341E18.2 |
-24.8kb | Anti-sense | Chr. 6 - 53057807 - 53100873 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

GCM1
Genomic Map