Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102723766 | LOC102723766 | -73.8kb | Sense | Chr. 4 - 184365561 - 184382306 on NCBI Build GRCh38 | |
| Downstream | liver cancer-associated transcript 8 | LVCAT8 | 162.0kb | Sense | Chr. 4 - 184584093 - 184624872 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map