Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105378146 | LOC105378146 | -69.0kb | Sense | Chr. 6 - 168927741 - 168964382 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101929460 | LOC101929460 | 121.0 | Sense | Chr. 6 - 169034171 - 169035643 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map