Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 362 | ZNF362 |
RN, lin-29 |
Sense | Chr. 1 - 33256307 - 33300719 on NCBI Build GRCh38 | |
| Downstream | alpha 1,3-galactosyltransferase 2 | A3GALT2 |
A3GALT2P, IGB3S, IGBS3S |
21.8kb | Anti-sense | Chr. 1 - 33306766 - 33321098 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map