Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LYR motif containing 7 | LYRM7 |
C5orf31, MC3DN8, MZM1L |
-159.8kb | Anti-sense | Chr. 5 - 131170914 - 131205426 on NCBI Build GRCh38 |
| Downstream | Rap guanine nucleotide exchange factor 6 | RAPGEF6 |
KIA001LB, PDZ-GEF2, PDZGEF2, RA-GEF-2, RAGEF2 |
58.3kb | Sense | Chr. 5 - 131423921 - 131635236 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map