Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Down syndrome critical region 8 | DSCR8 |
C21orf65, CT25.1a, CT25.1b, MMA-1, MMA-1a, MMA-1b, MMA1, MTAG2 |
-21.4kb | Sense | Chr. 21 - 38121451 - 38156511 on NCBI Build GRCh38 |
| Downstream | cutaneous T-cell lymphoma-associated antigen 1 pseudogene | LOC100288590 | 26.1kb | Anti-sense | Chr. 21 - 38204544 - 38205606 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map