Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101929319 | LOC101929319 | Anti-sense | Chr. 2 - 151337826 - 151379371 on NCBI Build GRCh38 | ||
| Downstream | microRNA 4773-1 | MIR4773-1 | 9.9kb | Sense | Chr. 2 - 151368334 - 151368411 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map