Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 717 | ZNF717 |
X17, ZNF838 |
-1.6mb | Anti-sense | Chr. 3 - 75729961 - 75785583 on NCBI Build GRCh38 |
| Downstream | roundabout guidance receptor 1 | ROBO1 |
DUTT1, SAX3 |
1.2mb | Anti-sense | Chr. 3 - 78597238 - 79767998 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map