Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
RNA, U105B small nucleolar |
RNU105B |
105B, E1-2 |
-32.9kb | Sense | Chr. 20 - 8831185 - 8831393 on NCBI Build GRCh38 |
| Downstream | phospholipase C beta 4 | PLCB4 |
ARCND2, PI-PLC |
201.7kb | Sense | Chr. 20 - 9068710 - 9480816 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map