Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PPP5 tetratricopeptide repeat domain containing 1 | PPP5D1 | Sense | Chr. 19 - 46480788 - 46601200 on NCBI Build GRCh38 | ||
| Downstream | calmodulin 3 | CALM3 |
HEL-S-72, PHKD, PHKD3 |
330.0 | Anti-sense | Chr. 19 - 46601255 - 46610782 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map