| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000315965.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000315965.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000315965.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000315965.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027073.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027073.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07027073.hg.1 |
TC07001740.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700187915.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187915.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187915.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187915.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187915.hg.1 |
TC0700012190.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700187914.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187914.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187914.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700187914.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700187914.hg.1 |
TC0700012190.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000315965.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000315965.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000315965.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000315965.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
ENST00000523505.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012190.hg.1 |
NM_175884.5 |
Refseq |