Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | family with sequence similarity 229 member B | FAM229B | C6orf225 | -114.9kb | Anti-sense | Chr. 6 - 112087471 - 112102790 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101927640 | LOC101927640 | 17.0kb | Anti-sense | Chr. 6 - 112236596 - 112306683 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map