Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC728095 | LOC728095 | -116.9kb | Anti-sense | Chr. 5 - 169013227 - 169037998 on NCBI Build GRCh38 | |
| Downstream | microRNA 585 | MIR585 |
MIRN585, hsa-mir-585, mir-585 |
107.4kb | Sense | Chr. 5 - 169263601 - 169263694 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map