Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | AFDN antisense RNA 1 (head to head) | AFDN-AS1 |
C6orf124, HGC6.4, MLLT4-AS1, dJ431P23.3 |
-72.9kb | Anti-sense | Chr. 6 - 167823890 - 167826796 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC100128124 | HGC6.3 | 74.1kb | Anti-sense | Chr. 6 - 167975924 - 167976939 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map