Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
synergin, gamma |
SYNRG |
AP1GBP1, SYNG |
Anti-sense | Chr. 17 - 37514797 - 37613637 on NCBI Build GRCh38 | |
| Downstream | DEAD-box helicase 52 | DDX52 |
HUSSY19, ROK1 |
57.6kb | Anti-sense | Chr. 17 - 37609739 - 37643464 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map